Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHIT Monoclonal Antibody (1093CT23.1.2)
GREENER_CHOICE

Catalog No. PIMA537675
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537675 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537675 Supplier Invitrogen™ Supplier No. MA537675
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FHIT Monoclonal Antibody for Western Blot, Flow

Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHIT
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1093CT23.1.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FHIT
Gene Accession No. P49789
Gene Alias AP3A hydrolase; AP3Aase; AW045638; bis(5'-adenosyl)-triphosphatase; bis5'-adenosyl-triphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; dinucleosidetriphosphatase; FHIT; Fra14A2; FRA3B; fragile histidine triad; fragile histidine triad gene; fragile histidine triad protein
Gene Symbols FHIT
Host Species Mouse
Immunogen FHIT recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.