Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL1 Antibody (OTI3G7), Janelia Fluor™ 669, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270739JF669
Description
FHL1 Monoclonal antibody specifically detects FHL1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/ImmunofluorescenceSpecifications
FHL1 | |
Monoclonal | |
Janelia Fluor 669 | |
bA535K18.1, four and a half LIM domains 1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1Isl-1 and Mec-3 domains 1 | |
Full length human recombinant protein of human FHL1 (NP_001440) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors | |
2273 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
OTI3G7 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction