Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL1 Antibody (OTI3G7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270739PCP
Description
FHL1 Monoclonal specifically detects FHL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| FHL1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| FHL1 | |
| Full length recombinant protein of human FHL1 (NP_001440) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI3G7 | |
| Flow Cytometry | |
| bA535K18.1, four and a half LIM domains 1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1Isl-1 and Mec-3 domains 1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors | |
| 2273.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction