Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL1 Antibody (SR1507), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321873100UL
Description
FHL1 Monoclonal antibody specifically detects FHL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
FHL1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR1507 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
bA535K18.1, four and a half LIM domains 1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1Isl-1 and Mec-3 domains 1 | |
A synthesized peptide derived from human FHL1 (Uniprot #: Q13642) | |
100 μg | |
Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors | |
2273 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction