Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518305 Supplier Invitrogen™ Supplier No. PA518305
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FHL1
Gene Accession No. P97447, Q9WUH4
Gene Alias bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1a; four and a half LIM domains 1b; four and a half LIM domains 1b; four and a half LIM domains protein 1b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1a; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBMX1A; RBMX1B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025
Gene Symbols FHL1
Host Species Goat
Immunogen Synthetic peptide sequence (CNKRFVFHNEQVY) corresponding to the internal amino acids of FHL1 (aa 261-272).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14199, 25177
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.