Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL1 Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA5142122

This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise isoforms 2 (NP_001440.1) and 5 (NP_001153171.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
FHL1 Polyclonal antibody specifically detects FHL1 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.
Specifications
FHL1 | |
Polyclonal | |
Biotin | |
FHL1 | |
bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1a; four and a half LIM domains 1b; four and a half LIM domains 1b; four and a half LIM domains protein 1b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1a; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBMX1A; RBMX1B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14199, 2273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P97447, Q13642 | |
FHL1 | |
Peptide with sequence CNKRFVFHNEQVY. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction