Learn More
FHL1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 109911AP150UL

Description
This is a rabbit polyclonal antibody raised against full length of human FHL1.
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.Specifications
FHL1 | |
Polyclonal | |
Unconjugated | |
FHL1 | |
FHL 1, FHL1, FHL1B, FLH1A, four and a half LIM domains 1, KYO T, Skeletal muscle LIM protein 1, SLIM, SLIM 1, SLIM1, XMPMA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14199, 2273, 387528, 399646 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P97447, Q13642 | |
FHL1, fhl1a, fhl1b | |
FHL1 Fusion Protein Ag1448 | |
150 μL | |
Primary | |
Human, Mouse, Zebrafish | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.