Learn More
Invitrogen™ FHL2 Monoclonal Antibody (F4B2-B11)

Description
MA1-40200 detects FHL2 from human, mouse, and rat samples. MA1-40200 has been successfully used in immunohistochemistry (frozen), immunohistochemistry (paraffin), Western blot, and immunofluorescence applications. The MA1-40200 immunogen is human FHL2. MA1-40200 recognizes the N-terminal Zn-Finger motif, it does not crossreact in Western Blotting with the FHL1 and FHL3 proteins. MA1-40200 antibody is cross reactive with mouse and rat FHL2.
Specifications
Specifications
| Antigen | FHL2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | F4B2-B11 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | Fhl2 |
| Gene Accession No. | O35115, O70433, Q14192 |
| Gene Alias | AAG11; aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; DRAL; Fhl2; FHL-2; four and a half LIM domains 2; four and a half LIM domains protein 2; LIM domain protein DRAL; OTTHUMP00000203164; OTTHUMP00000203168; OTTHUMP00000203169; skeletal muscle LIM-protein 3; SLIM 3; SLIM3; SLIM-3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.