Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582232 Supplier Invitrogen™ Supplier No. PA582232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFDCHHCNES LFGKKYILRE ESPYCVVCFE TLFANTCEEC GKPIGCDCKD LSYKDRHWHE ACFHCSQCRN SLVDKPFAAK EDQLLCTDCY SNEYSSKCQE CKKTIMPGTR KMEYKGSS Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. This protein is thought to have a role in the assembly of extracellular membranes. Also, this gene is down-regulated during transformation of normal myoblasts to rhabdomyosarcoma cells and the encoded protein may function as a link between presenilin-2 and an intracellular signaling pathway. Multiple alternatively spliced variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fhl2
Gene Accession No. O35115, O70433, Q14192
Gene Alias AAG11; aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; DRAL; Fhl2; FHL-2; four and a half LIM domains 2; four and a half LIM domains protein 2; LIM domain protein DRAL; OTTHUMP00000203164; OTTHUMP00000203168; OTTHUMP00000203169; skeletal muscle LIM-protein 3; SLIM 3; SLIM3; SLIM-3
Gene Symbols Fhl2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FHL2. Recombinant protein control fragment (Product #RP-100978).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14200, 2274, 63839
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.