Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL2 Rabbit anti-Human, Mouse, Rat, Clone: 0K9C7, Novus Biologicals™

Rabbit Monoclonal Antibody
$196.25 - $452.15
Specifications
| Antigen | FHL2 |
|---|---|
| Clone | 0K9C7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
FHL2 Monoclonal antibody specifically detects FHL2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FHL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2274 | |
| IgG | |
| Affinity purified |
| 0K9C7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-260 of human FHL2 (Q14192). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title