Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FHL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531181
Description
Recommended positive controls: HeLa, HCT116. Predicted reactivity: Mouse (96%), Rat (92%), Pig (97%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.
Specifications
| FHL3 | |
| Polyclonal | |
| Unconjugated | |
| FHL3 | |
| FHL3; FHL-3; four and a half LIM domains 3; four and a half LIM domains protein 3; LIM-only protein FHL3; MGC23614; MGC8696; skeletal muscle LIM-protein 2; SLIM2; SLIM-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2275 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.99 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q13643 | |
| FHL3 | |
| Recombinant protein encompassing a sequence within the center region of human FHL3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction