Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595379
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595379 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595379 Supplier Invitrogen™ Supplier No. PA595379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, rat heart tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL3
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene FHL3
Gene Accession No. Q13643
Gene Alias FHL3; FHL-3; four and a half LIM domains 3; four and a half LIM domains protein 3; LIM-only protein FHL3; MGC23614; MGC8696; skeletal muscle LIM-protein 2; SLIM2; SLIM-2
Gene Symbols FHL3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human FHL3 (41-84aa AECQQLIGHDSRELFYEDRHFHEGCFRCCRCQRSLADEPFTCQD).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2275, 313582
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.