Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597144 Supplier Invitrogen™ Supplier No. PA597144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSESFDCAKC NESLYGRKYI QTDSGPYCVP CYDNTFANTC AECQQLIGHD SRELFYEDRH FHEGCFRCCR CQRSLADEPF TCQDSELLCN DCYCSAFSSQ CSACGETVMP GSRKLEYGGQ TWHEHCFLCS GCEQPLGSRS FVPDKGAHYC VPCYENKFAP RCARCSKTLT QGGVTYRDQP WHRECLVCTG CQTPLAGQQF TSRDEDPYCV ACFGELFAPK CSSCKRPIVG LGGGKYVSFE DRHWHHNCFS CARCSTSLVG QGFVPDGDQV LCQGCSQAGP; Positive Samples: LO2, HT-1080, HL-60, Mouse skeletal muscle.

LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FHL3
Gene Accession No. Q13643, Q9R059
Gene Alias FHL3; FHL-3; four and a half LIM domains 3; four and a half LIM domains protein 3; LIM-only protein FHL3; MGC23614; MGC8696; skeletal muscle LIM-protein 2; SLIM2; SLIM-2
Gene Symbols FHL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human FHL3 (NP_0044592).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14201, 2275
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.