Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304622100UL
Description
FHL3 Polyclonal specifically detects FHL3 in Human, Mouse samples. It is validated for Western Blot.Specifications
FHL3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2275 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FHL-3, four and a half LIM domains 3, four and a half LIM domains protein 3, LIM-only protein FHL3, MGC19547, MGC8696, Skeletal muscle LIM-protein 2, SLIM-2, SLIM2MGC23614 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human FHL3 (NP_004459.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction