Learn More
FHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 110282AP150UL

Description
This antibody recognizes specifically the 34 kDa FHL3 protein.
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.Specifications
FHL3 | |
Polyclonal | |
Unconjugated | |
FHL3 | |
FHL 3, FHL3, four and a half LIM domains 3, Skeletal muscle LIM protein 2, SLIM 2, SLIM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14201, 2275, 313582 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13643, Q9R059 | |
Fhl3 | |
FHL3 Fusion Protein Ag1495 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.