Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FHL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521814
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (89%), Rat (90%), Pig (92%), Chimpanzee (98%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
FHL5 | |
Polyclonal | |
Unconjugated | |
FHL5 | |
1700027G07Rik; Act; activator of cAMP-responsive element modulator (CREM) in testis; activator of cAMP-responsive element modulator in testis; Activator of CREM in testis; dJ393D12.2; FHL5; FHL-5; four and a half LIM domains 5; four and a half LIM domains protein 5; LIM protein ACT; LIM-only protein ACT; RP3-393D12.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9457 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q5TD97 | |
FHL5 | |
Recombinant protein encompassing a sequence within the center region of human FHL5. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction