Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113947 Supplier Invitrogen™ Supplier No. PA5113947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIMPGSRKME FKGNYWHETC FVCENCRQPI GTKPLISKES GNYCVPCFEK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FHL5
Gene Accession No. Q5TD97
Gene Alias 1700027G07Rik; Act; activator of cAMP-responsive element modulator (CREM) in testis; activator of cAMP-responsive element modulator in testis; Activator of CREM in testis; dJ393D12.2; FHL5; FHL-5; four and a half LIM domains 5; four and a half LIM domains protein 5; LIM protein ACT; LIM-only protein ACT; RP3-393D12.2
Gene Symbols FHL5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FHL5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.