Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHOD1 Monoclonal Antibody (M352)
GREENER_CHOICE

Catalog No. PIMA547717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547717 Supplier Invitrogen™ Supplier No. MA547717
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 140 kDa* protein corresponding to the apparent molecular mass of FHOD1 on SDS-PAGE immunoblots of human K562 and Jurkat, and has weak reactivity to FHOD1 in mouse C2C12.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHOD1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone M352
Concentration 0.217 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene FHOD1
Gene Accession No. Q9Y613
Gene Alias BC017144; FH1/FH2 domain-containing protein 1; Fhod1; FHOS; FHOS1; formin homolog overexpressed in spleen 1; formin homology 2 domain containing 1; formin homology 2 domain-containing protein 1
Gene Symbols FHOD1
Host Species Mouse
Immunogen FHOD1 recombinant protein that included the full length human FHOD1 sequence. This sequence has 50% homology to human FHOD3, and low homology to other formin family members.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29109
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.