Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115233 Supplier Invitrogen™ Supplier No. PA5115233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FHOD1.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHOD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FHOD1
Gene Accession No. Q6P9Q4, Q9Y613
Gene Alias BC017144; FH1/FH2 domain-containing protein 1; Fhod1; FHOS; FHOS1; formin homolog overexpressed in spleen 1; formin homology 2 domain containing 1; formin homology 2 domain-containing protein 1
Gene Symbols FHOD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FHOD1(Accession Q9Y613), corresponding to amino acid residues L336-A386.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234686, 29109, 291964
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.