Learn More
FHOD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304825A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function of this protein remains unknown.Specifications
FHOD1 | |
Polyclonal | |
Unconjugated | |
FHOD1 | |
FHOS, formin homolog overexpressed in spleen 1, Formin homology 2 domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29109 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y613 | |
FHOD1 | |
Between 1114 and 1164 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.