Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHOD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30437320UL
Description
FHOD1 Polyclonal specifically detects FHOD1 in Human samples. It is validated for Western Blot.Specifications
FHOD1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
FH1/FH2 domain-containing protein 1, FHOS1, FHOSFormin homolog overexpressed in spleen 1, formin homology 2 domain containing 1, Formin homology 2 domain-containing protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1010-1120 of human FHOD1 (NP_037373.2). | |
20 μL | |
Apoptosis | |
29109 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction