Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FHOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555158
Description
Immunogen sequence: QCGDILTNKR FMLDMLYAHN RKSPDDEEKG DGEAGRTQQE AEAVASLATR ISTLQANSQT QDESVRRVDV GCLDNRGSVK AFAEKFNSGD LGRGSISPDA EPNDKVPETA PVQPKTESDY IWDQLM Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.
FHOD3 is a novel member of the formin homology family playing an important role in regulation of the actin cytoskeleton. It contains a DAD (diaphanous auto regulatory) domain, an FH1 (formin homology 1) domain, an FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. FHOD3 is a cytoplasmic protein where the main part of the protein localizes to actin fibers and the remaining part displays filamentous staining. A 1422 amino acid containing formin-related actin-organizing protein, FHOD3 may cause stress fiber formation together with cell elongation and may associate with nestin/NES-based interfilament (IF). Expression of FHOD3 is seen mainly in heart, kidney, and brain.
Specifications
| FHOD3 | |
| Polyclonal | |
| Unconjugated | |
| FHOD3 | |
| A930009H06Rik; FH1/FH2 domain-containing protein 3; Fhod3; Fhos2; Formactin2; Formactin-2; Formin homolog overexpressed in spleen 2; formin homology 2 domain containing 3; formin-family protein FHOS2; hFHOS2; KIAA1695; mFHOS2; mKIAA1695 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80206 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2V2M9 | |
| FHOD3 | |
| Recombinant protein corresponding to Human FHOD3. Recombinant protein control fragment (Product #RP-88592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction