Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523313
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523313 Supplier Invitrogen™ Supplier No. PA523313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FHOD3 Polyclonal Antibody for IHC (P)

FHOD3 is a novel member of the formin homology family playing an important role in regulation of the actin cytoskeleton. It contains a DAD (diaphanous auto regulatory) domain, an FH1 (formin homology 1) domain, an FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. FHOD3 is a cytoplasmic protein where the main part of the protein localizes to actin fibers and the remaining part displays filamentous staining. A 1422 amino acid containing formin-related actin-organizing protein, FHOD3 may cause stress fiber formation together with cell elongation and may associate with nestin/NES-based interfilament (IF). Expression of FHOD3 is seen mainly in heart, kidney, and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHOD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene FHOD3
Gene Accession No. Q2V2M9, Q76LL6
Gene Alias A930009H06Rik; FH1/FH2 domain-containing protein 3; Fhod3; Fhos2; Formactin2; Formactin-2; Formin homolog overexpressed in spleen 2; formin homology 2 domain containing 3; formin-family protein FHOS2; hFHOS2; KIAA1695; mFHOS2; mKIAA1695
Gene Symbols FHOD3
Host Species Rabbit
Immunogen A portion of amino acids 1250-1300 of human FHOD3 (CRA-d).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100067548, 225288, 420952, 480157, 785433, 80206
Target Species Human, Mouse, Canine, Bovine, Horse, Chicken, Marsupial
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.