Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIBCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543502 Supplier Invitrogen™ Supplier No. PA543502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DGYPLTVADY SGTAGDSLLK HSGMRFTTKD RDSDHSENNC AAFYRGAWWY Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

FIBCD1 is a single-pass membrane protein. It contains 1 fibrinogen C-terminal domain. The exact function of FIBCD1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIBCD1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FIBCD1
Gene Accession No. A2AV25, Q8N539
Gene Alias AI448887; Fibcd1; fibrinogen C domain containing 1; fibrinogen C domain-containing protein 1; fibrinogen C domain-containing protein 1; uncharacterized protein LOC567525; LOW QUALITY PROTEIN: fibrinogen C domain-containing protein 1; RGD1309097; si:dkey-54o1.1; UNQ701/PRO1346
Gene Symbols FIBCD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FIBCD1 (aa 362-411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84929, 98970
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.