Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Catalog No. 13994081
Change view
Click to view available options
Holds:
100 (10 x 10 Grid)
16 (4 x 4 Grid)
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Quantity
13-994-081 16 (4 x 4 Grid) Each
11-678-24C 100 (10 x 10 Grid) Each
11-678-26D 49 (7 x 7 Grid) 12 Pack
13-992-33 64 (8 x 8 Grid) 12 Pack
13-989-218 81 (9 x 9 Grid) 12 Pack
13-994-085 25 (5 x 5 Grid) Each
13-994-091 49 (7 x 7 Grid) Each
13-994-092 64 (8 x 8 Grid) Each
13-994-104 100 (10 x 10 Grid) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-994-081 Supplier Thermo Scientific™ Supplier No. 820016
Add to Cart
Add to Cart
Organize vial storage within cryo boxes with these fiberboard box dividers.
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Holds 16 (4 x 4 Grid)