Learn More
Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
GREENER_CHOICE
Catalog No. p-4528008
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Conduct high-volume microplate processing with this large volume swinging bucket rotor.
Conduct high-volume microplate processing with the Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor.
Features:
Fiberlite rotors:
  • Lightweight carbon fiber rotors reduce run-times by achieving higher G-forces
  • Corrosion-and fatigue-resistant design ensures structural integrity
  • Rotors are backed by a 15-year warranty and are compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Recommended for:
Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite H3-LV Large Volume Swinging Bucket Rotor for Thermo Scientific Sorvall Legend XT, Multifuge X3 Series
Type Swinging Bucket Rotor
Capacity 28 Standard or 8 Deepwell Plates
Max. RCF 1,840 x g
Max. Speed 3,600 rpm
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle 90°
Biocontainment Yes
Green Features Energy efficient, less waste
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.