Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIBIN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983949
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3949 Supplier Bioss Supplier No. BS16085R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen FIBIN
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FIBIN
Gene Accession No. Q8TAL6
Gene Alias FIBIN; Fin bud initiation factor homolog; fin bud initiation factor homolog (zebrafish); PSEC0235
Gene Symbols FIBIN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FIBIN.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387758
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.