Learn More
Invitrogen™ FIBP Monoclonal Antibody (1E5)

Description
Peptide Sequence: MTSELDIFVG NTTLIDEDVY RLWLDGYSVT DAVALRVRSG ILEQTGATAA VLQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGIT LKSCRRQFDN FKRVFKVVEE MRGSLVDNIQ QHFLLSDRLA RDYAAIVFFA NNRFETGKKK LQYLSFGDFA FCAELMIQNW TLGAVDSQMD DMDMDLDKEF LQDLKELKVL VADKDLLDLH KSLVCTALRG KLGVFSEMEA NFKNLSRGLV NVAAKLTHNK DVGDLFVDLV EKFVEPCRSD HWPLSDVRFF LNQYSASVHS LDGFRHQALW DRYMGTLRGC LLRLYHD.
Specifications
Specifications
| Antigen | FIBP |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1E5 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FIBP |
| Gene Accession No. | O43427 |
| Gene Alias | 2010004G08Rik; 2010005N21Rik; 3010027N18Rik; Acidic fibroblast growth factor intracellular-binding protein; aFGF intracellular binding protein; aFGF intracellular-binding protein; FGF intracellular binding protein; FGF1 intracellular binding protein; FGF-1 intracellular-binding protein; FGFIBP; Fibp; FIBP-1; fibroblast growth factor (acidic) intracellular binding protein |
| Gene Symbols | FIBP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.