Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIBP Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589306 Supplier Thermo Scientific Supplier No. PA589306

Rabbit Polyclonal Antibody

Immunogen sequence: MTSELDIFVG NTTLIDEDVY RLWLDGYSVT DAVALRVRSG ILEQTGATAA VLQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGIT LKSCRRQFDN FKRVFKVVEE MRGSLVDNIQ QHFLLSDRLA RDYAAIVFFA NNRFETGKKK LQYLSFGDFA FCAELMIQNW TLGAVDSQMD DMDMDLDKEF LQDLKELKVL VADKDLLDLH KSLVCTALRG KLGVFSEMEA NFKNLSRGLV NVAAKLTHNK DVRDLFVDLV EKFVEPCRSD HWPLSDVRFF LNQYSASVHS LDGFRHQALW DRYMGTLRGC LLRLYHD; Positive Samples: BXPC-3, Mouse skeletal muscle; Cellular Location: Endomembrane system, Nucleus, Peripheral membrane protein

Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene FIBP
Gene Accession No. O43427, Q9JI19
Gene Alias 2010004G08Rik, 2010005N21Rik, 3010027N18Rik, Acidic fibroblast growth factor intracellular-binding protein, aFGF intracellular binding protein, aFGF intracellular-binding protein, FGF intracellular binding protein, FGF1 intracellular binding protein, FGF-1 intracellular-binding protein, FGFIBP, Fibp, FIBP-1, fibroblast growth factor (acidic) intracellular binding protein
Gene Symbols FIBP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human FIBP (NP_0042052)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58249, 9158
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.