Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIBP Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30470120UL
Description
FIBP Polyclonal specifically detects FIBP in Human, Rat samples. It is validated for Western Blot.Specifications
FIBP | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
Unconjugated | |
fibroblast growth factor (acidic) intracellular binding protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human FIBP (NP_004205.2). | |
20 μL | |
Signal Transduction | |
9158 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction