Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Fibre-Metal™ Face Shield Headgear

Catalog No. 19085618 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Includes:
Crown with Mounting Cups
Faceshield Adaptor
Headgear Only
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-085-618 Crown with Mounting Cups
19-085-619 Faceshield Adaptor
19-017853 Headgear Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-618 Supplier Honeywell™ Supplier No. F4400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal for secure, long-term combination use

  • Window is not included and sold separately
  • Quick-Lok™ Cups can be can be securely mounted onto, and removed from the cap without tools

Specifications

Width (English) 4.5 in.
Certifications/Compliance ANSI Z87.1
Color Gray
Product Type Headgear
For Use With (Equipment) Attaches to E2Q, P2HNQ Series Hard Hats
Dimensions (L x W x H) 10.2 x 4.5 x 10.12 in.
Includes Crown with Mounting Cups
No. per Pack 1/Pk.
Style Partial Crown
Weight (English) 7.2 oz.
Material Noryl™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.