Learn More
Honeywell™ Fibre-Metal™ Supereight™ 3-R Ratchet Headband
Shop All Protective Industrial Products Inc. ProductsSpecifications
Specifications
| Certifications/Compliance | ANSI-Z89.1 TYPE 1, Class E, G, or C |
| Color | Hi-vis Orange |
| Finish | Smooth |
| Material | Superlectric Thermoplastic |
| Quantity | 1 Ea. |
| Suspension Type | 8-point 3R Rachet |
| Head Style | Smooth Crown |