Learn More
Honeywell™ Fibre-Metal™ Tigerhood™ Classic Welding Helmets
Catalog No. 19017939 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Welding Helmet (Passive)
Welding Helmet w/loop mounting system
Welding Helmet with Mounting Cups
Style:
Passive design
With loop mounting system
With mounting cups
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Style
19-017939 Welding Helmet with Mounting Cups With mounting cups
17-986-523 Welding Helmet w/loop mounting system With loop mounting system
17-986-522 Welding Helmet (Passive) Passive design
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-017939 Supplier Honeywell™ Supplier No. 4906GY
  • $163.33
    $115.65 / Each of 1
    Save $47.68
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Provide extended throat protection while blocking fumes and smoke from face
  • Molded-in color eliminates chipping, peeling and fading
  • Smooth, seamless shell deflects sparks and spatter
  • Thermoplastic
  • Lift front
Specifications
Color Gray
Includes Mounting cups
Material Thermoplastic
Product Type Welding Helmet with Mounting Cups
Style With mounting cups