Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Fibre-Metal™ Visors for F400 Headgear

Catalog No. 19017927 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Height (English):
8 in.
8.75 in.
Lens Tint:
Clear
Shade 5
Width (English):
16.5 in.
19 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Lens Tint Width (English)
19-017927 8 in. Clear 16.5 in.
19-017929 8.75 in. Clear 19 in.
19-811-502 8 in. Shade 5 16.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-017927 Supplier Honeywell™ Supplier No. 4178CL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Height (English) 8 in.
Width (English) 16.5 in.
Lens Tint Clear
For Use With (Equipment) F400 headgear in.
Lens Material Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.