Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillarin Antibody, Novus Biologicals™
SDP

Catalog No. p-200006078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB237245 100 μg
NB237585 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB237245 Supplier Novus Biologicals Supplier No. NBP321414100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fibrillarin Polyclonal antibody specifically detects Fibrillarin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Fibrillarin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: EYRAWNPFRSKLAAAILGGVDQIHIKP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2091
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.