Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrillarin Monoclonal Antibody (38F3)
GREENER_CHOICE

Catalog No. AMSA6771
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMSA6771 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMSA6771 Supplier Invitrogen™ Supplier No. MA316771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: antigen standard for FBL (transient overexpression lysate).

Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillarin
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 38F3
Concentration Conc. not determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.035% sodium azide
Gene fbl
Gene Accession No. P15646, P22087, P22509, P35550, Q22053
Gene Alias 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7
Gene Symbols fbl, fib-1, Nop1p
Host Species Mouse
Immunogen Yeast nuclear preparation (S. cerevisiae).
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100064571, 100626002, 14113, 179999, 2091, 292747, 46234, 538548, 851548
Target Species Bovine, C. elegans, Chicken, Drosophila, Horse, Human, Mouse, Plant, Pig, Rat, Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.