Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillarin Mouse anti-Mouse, Yeast, Drosophila, Rat, Human, Clone: 38F3, MilliporeSigma™

Catalog No. MABE1154MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1154MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1154MI Supplier MilliporeSigma Supplier No. MABE1154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Fibrillarin, clone 38F3 is a highly specific mouse monoclonal antibody that targets Fibrillarin/Nop1p and has been tested in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillarin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 38F3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols NOP1,LOT3,YDL014W,D2870
Host Species Mouse
Immunogen Yeast nuclear extract
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_010270.1
Test Specificity Clone 38F3 specifically detects Fibrillarin/Nop1p in multiple mammalian species and in yeast.
Target Species Mouse, Yeast, Drosophila, Rat, Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.