Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrillarin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109251 Supplier Invitrogen™ Supplier No. PA5109251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Hela, HepG2, 293T, NIH-3T3, HL-60.

Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillarin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene fbl
Gene Accession No. P22087
Gene Alias 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7
Gene Symbols fbl
Host Species Rabbit
Immunogen Synthetic peptide within Human Fibrillarin aa 272-321.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2091
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.