Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fibrillarin Polyclonal Antibody
Chicken Polyclonal Antibody
$514.50 - $1763.00
Specifications
Antigen | Fibrillarin |
---|---|
Concentration | Conc. Not Determined |
Content And Storage | 4°C |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143531
|
Invitrogen™
PA5143531 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143565
|
Invitrogen™
PA5143565 |
100 μL |
Each for $514.50
|
|
|||||
Description
Fibrillarin Polyclonal Antibody for Western Blot, ICC/IF
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).Specifications
Fibrillarin | |
4°C | |
Polyclonal | |
Liquid | |
IgY | |
Canine, Horse, Human, Mouse, Rat | |
fbl | |
34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7 | |
fbl | |
Primary |
Conc. Not Determined | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Chicken | |
RUO | |
PBS with 0.02% sodium azide | |
P22087, P22509, P35550 | |
100064571, 14113, 2091, 292747, 476460 | |
Full length human fibrillarin expressed in and purified from E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title