Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillarin Rabbit anti-Human, Mouse, Rat, Clone: 9J5O2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Fibrillarin |
---|---|
Clone | 9J5O2 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Fibrillarin Monoclonal antibody specifically detects Fibrillarin in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Fibrillarin | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2091 | |
IgG | |
Affinity purified |
9J5O2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
A synthetic peptide corresponding to a sequence within amino acids 222-321 of human Fibrillarin (P22087). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title