Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillin 1 Antibody (FBN1/6933), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320532100UG
Description
Fibrillin 1 Monoclonal antibody specifically detects Fibrillin 1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Fibrillin 1 | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Asprosin, FBN, fibrillin 1, fibrillin 1 (Marfan syndrome), fibrillin 15, fibrillin-1, MASS, MFS1, OCTD, SGS, SSKS, WMS | |
Recombinant human Fibrillin 1 fragment around aa 330-476 (Exact sequence is proprietary) | |
100 μg | |
Extracellular Matrix | |
2200 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
FBN1/6933 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction