Learn More
Invitrogen™ Fibrillin 2 Monoclonal Antibody (1C2)

Description
Peptide Sequence: RQKRSIHEPD PTAVEQISLE SVDMDSPVNM KFNLSHLGSK EHILELRPAI QPLNNHIRYV ISQGNDDSVF RIHQRNGLSY LHTAKKKLMP GTYTLEITSI P.
Specifications
Specifications
| Antigen | Fibrillin 2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FBN2 |
| Gene Accession No. | P35556 |
| Gene Alias | BC063774; CCA; DA9; EOMD; Fbn2; Fbn-2; Fib-2; fibrillin 2; fibrillin 5; fibrillin-2; Fibrillin-2 C-terminal peptide; mKIAA4226; sne; sy; syndatyly ems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.