Learn More
fibrillin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 202521AP150UL

Description
This antibody is specific to FBN2. It recognizes both two isoforms ∽300 kDa and 160 kDa.
The protein encoded by this gene is a component of connective tissue microfibrils and may be involved in elastic fiber assembly. Mutations in this gene cause congenital contractural arachnodactyly.Specifications
fibrillin 2 | |
Polyclonal | |
Unconjugated | |
FBN2 | |
CCA, DA9, FBN2, fibrillin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14119, 2201, 689008 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35556, Q61555 | |
Fbn2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.