Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrin Antibody (UC45), Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Fibrin |
---|---|
Clone | UC45 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF |
Description
Fibrin Monoclonal antibody specifically detects Fibrin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays, CyTOFSpecifications
Fibrin | |
1 mg/mL | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Biology | |
PBS | |
IgM | |
Thiophilic Chromatography |
UC45 | |
Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Human acute monoblastic leukemia cells. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title