Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrin beta-chain Antibody (8E5), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281111MFV500
Description
Fibrin beta-chain Monoclonal antibody specifically detects Fibrin beta-chain in Human samples. It is validated for Flow CytometrySpecifications
Fibrin beta-chain | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
800000 | |
Flow Cytometry | |
Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p | |
8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin). | |
0.1 mL | |
Cell Cycle and Replication | |
2244 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction