Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrin Mouse anti-Human,Mouse, KHL, Clone: 59D8, MilliporeSigma™

Catalog No. MABS21551MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21551MI 100 μg
MABS21552MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21551MI Supplier MilliporeSigma Supplier No. MABS2155100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrin Monoclonal antibody specifically detects Fibrin in Human,Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry, Peptide Inhibition Assay, Radioimmunoassay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrin
Applications Immunofluorescence, Immunohistochemistry, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 59D8
Conjugate KLH
Gene Accession No. NP_001171670
Gene Alias Fibrinogen beta chain
Gene Symbols FGB
Host Species Mouse
Immunogen KLH-conjugated synthetic heptapeptide (GHRPLNK) derived from the N-terminal region of the beta chain of human fibrinogen.
Molecular Weight of Antigen ∼55 kDa observed; 55.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 59D8 is a mouse monoclonal antibody that specifically detects human and murine fibrin. It targets an epitope within a heptapeptide fom the N-terminal region of the beta chain.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.