Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrinogen alpha chain Monoclonal Antibody (471CT12.1.2)
GREENER_CHOICE

Catalog No. PIMA537571
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537571 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537571 Supplier Invitrogen™ Supplier No. MA537571
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrinogen alpha chain Monoclonal Antibody for Western Blot

Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinogen alpha chain
Applications Western Blot
Classification Monoclonal
Clone 471CT12.1.2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Fga
Gene Accession No. E9PV24, P02671
Gene Alias Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen a chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogena chain; Fibrinopeptide A
Gene Symbols Fga
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 420-448 amino acids from the N-terminal region of human FGA.
Purification Method Euglobulin precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14161, 2243
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.