Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrinogen alpha chain Recombinant Rabbit Monoclonal Antibody (7K8J1)
GREENER_CHOICE

Catalog No. PIMA537943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537943 Supplier Invitrogen™ Supplier No. MA537943
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Human plasma, Rat liver Immunogen sequence: NDEGEGEFWL GNDYLHLLTQ RGSVLRVELE DWAGNEAYAE YHFRVGSEAE GYALQVSSYE GTAGDALIEG SVEEGAEYTS HNNMQFSTFD RDADQWEENC A.

Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinogen alpha chain
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7K8J1
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fga
Gene Accession No. P06399
Gene Alias Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain; Fibrinopeptide A
Gene Symbols Fga
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361969
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.