Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen Antibody [Allophycocyanin], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4786A
Description
Fibrinogen Polyclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Western BlotSpecifications
Fibrinogen | |
Polyclonal | |
PBS | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
2243 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
APC | |
Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
Human serum-derived Fibrinogen | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction