Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fibrinogen, Bovine

Catalog No. AAJ6327609
Click to view available options
Quantity:
1g
5g
10g
25g

Fibrinogen (CAS # 9001-32-5) is a glycoprotein produced in the vertebrate liver that aids in clot formation.

A soluble plasma glycoprotein, produced by the liver, that is converted by thrombin into fibrin during the blood coagulation process. Fibrinogen is a natural blood protein involved in the coagulation process. It is used instead of traditional sources of fibrinogen such as the blood products, fresh frozen plasma and cryoprecipitate. It helps in blood clot formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fibrinogen exists in blood as a dimer of three bonded, non-identical chains, FGA, FGG, and FGB

• Fibrinogen is cleaved by thrombin to form fibrin, the most abundant component of blood clots

Application

• Fibrinogen can be used in a laboratory setting to form polymer hydrogels, particularly for use as a tissue scaffold

• Fibrinogen may be useful in the research of neurodegeneration and brain injury; some evidence shows that it has a signaling role in neural progenitor cells

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Neural Signaling
Formulation Lyophilized powder
Quantity 10g
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Fibrinogen, Bovine
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.